Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RHCE Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113750
Description
Immunogen sequence: SSKYPRSVRR CLPLCALTLE AALILLFYFF THYDASLEDQ KGLVASYQVG For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Human: 100%.
May be part of an oligomeric complex which is likely to have a transport or channel function in the erythrocyte membrane.
Specifications
| RHCE | |
| Polyclonal | |
| Unconjugated | |
| RHCE | |
| (C)ces type 1 Rhesus blood group D antigen; blood group Rh(CE) polypeptide; blood group RhCcEe antigen; blood group RhCE polypeptide; CD240CE; RH; Rh blood group antigen Evans; Rh blood group C antigen; Rh blood group CcEe antigens; Rh blood group protein; Rh blood group, CcEe antigens; rh polypeptide 1; Rh polypeptide I; Rh30A; Rh4; RHC; RHCE; RhCE blood group antigens; RHCE blood group variant Crawford antigen Rh43; RHE; rhesus blood group antigen, RhC antigen; Rhesus blood group CE protein; Rhesus blood group E antigen; rhesus blood group little e antigen; Rhesus blood group Rhce antigen; rhesus C/E antigens; Rhesus system C and E polypeptides; RhIVb(J); RHIXB; RhPI; RhVI; RhVIII; silenced Rh blood group CcEe antigen; truncated RhCcEe antigen; truncated RHCE | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 6006 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P18577 | |
| RHCE | |
| Synthetic peptide directed towards the N terminal region of human RHCE. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction