Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RHCE Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571360
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571360 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571360 Supplier Thermo Scientific Supplier No. PA571360
Add to Cart
Add to Cart

RHCE Polyclonal Antibody

RHCE Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen RHCE
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RHCE
Gene Accession No. P18577
Gene Alias (C)ces type 1 Rhesus blood group D antigen; blood group Rh(CE) polypeptide; blood group RhCcEe antigen; blood group RhCE polypeptide; CD240CE; RH; Rh blood group antigen Evans; Rh blood group C antigen; Rh blood group CcEe antigens; Rh blood group protein; Rh blood group, CcEe antigens; rh polypeptide 1; Rh polypeptide I; Rh30A; Rh4; RHC; RHCE; RhCE blood group antigens; RHCE blood group variant Crawford antigen Rh43; RHE; rhesus blood group antigen, RhC antigen; Rhesus blood group CE protein; Rhesus blood group E antigen; rhesus blood group little e antigen; Rhesus blood group Rhce antigen; rhesus C/E antigens; Rhesus system C and E polypeptides; RhIVb(J); RHIXB; RhPI; RhVI; RhVIII; silenced Rh blood group CcEe antigen; truncated RhCcEe antigen; truncated RHCE
Gene Symbols RHCE
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human RHCE.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6006
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.