Learn More
Invitrogen™ RHCG Polyclonal Antibody

Description
The antibody detects endogenous levels of total RHCG protein.
Specifications
Specifications
| Antigen | RHCG |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 2.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | RHCG |
| Gene Accession No. | Q9UBD6 |
| Gene Alias | ammonium transporter Rh type C; BB065800; C15orf6; CDRC2; PDRC2; Rh family; Rh family C glycoprotein; rh family type C glycoprotein; Rh family, C glycoprotein; rh glycoprotein kidney; Rh type C glycoprotein; RHCG; Rhesus blood group family type C glycoprotein; Rhesus blood group, C glycoprotein; Rhesus blood group-associated C glycoprotein; Rhesus-associated C glycoprotein; RHGK; SLC42A3; Tumor-related protein DRC2 |
| Gene Symbols | RHCG |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.