Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RHCG Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560320
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560320 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560320 Supplier Invitrogen™ Supplier No. PA560320
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RYDFEADAHW WSERTHKNLS DMENEFYYRY P Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 72%.

RHCG (Rhesus blood group family type C glycoprotein), also known as RHGK (Rh glycoprotein kidney), PDRC2 (tumor-related protein DRC2) or SLC42A3, is a multi-pass membrane protein with three potential N-glycosylation sites that belongs to the Rh subfamily of the ammonium transporter family. Expressed in a wide variety of tissues with predominant expression in adult and fetal kidney, RHCg localizes to the apical and/or basolateral cell membrane (depending on the species) and is believed to function as a non-erythroid, bidirectional, electroneutral ammonium transporter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RHCG
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RHCG
Gene Accession No. Q9UBD6
Gene Alias ammonium transporter Rh type C; BB065800; C15orf6; CDRC2; PDRC2; Rh family; Rh family C glycoprotein; rh family type C glycoprotein; Rh family, C glycoprotein; rh glycoprotein kidney; Rh type C glycoprotein; RHCG; Rhesus blood group family type C glycoprotein; Rhesus blood group, C glycoprotein; Rhesus blood group-associated C glycoprotein; Rhesus-associated C glycoprotein; RHGK; SLC42A3; Tumor-related protein DRC2
Gene Symbols RHCG
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RHCG. Recombinant protein control fragment (Product #RP-98469).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51458
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.