Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RHD Monoclonal Antibody (C1), Invitrogen™

Catalog No. FEMA551186
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA551186 20 μL
FEMA551185 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEMA551186 Supplier Invitrogen™ Supplier No. MA551186
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The Rhesus (Rh) blood group system represents one of the most complex and important systems in humans. Two highly homologous genes RHD and RHCE (collectively referred to as RH30 or RHCED) encode the antigens of the Rh blood group system (1-4). These tightly linked genes map to human chromosomal position 1p34.1-1p36. The RHD gene, which is commonly deleted from a large segment of the population, encodes the most potent blood group immunogen, the D antigen (2,5). Rh incompatibility between maternal and fetal blood types results in hemolytic disease of the newborn (HDN), which often results in fetal death (1,2,6). The RHCE gene exists in four allelic forms, and each allele determines the expression of two antigens in Ce, ce, cE, or CE combinations (2,4,5). The RHCED antigens exist as integral membrane proteins with contain 12-transmembrane helices, and maintain erythrocyte membrane integrity. The presentation of the Rh antigenic activity requires the formation of a complex between the RHCED antigens and RHAG (RH50).
TRUSTED_SUSTAINABILITY

Specifications

Antigen RHD
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone C1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene RHD
Gene Accession No. Q02161
Gene Alias blood group antigen D; blood group protein RHD; blood group Rh(D) polypeptide; CD240D; D antigen (DCS); DIIIc; Erythrocyte membrane glycoprotein Rh30; Rh; Rh blood group antigen Evans; Rh blood group D antigen; Rh blood group protein; Rh blood group, D anitgen; Rh blood group, D antigen; RH polypeptide 2; Rh30; Rh4; Rhced; RHD; RhD blood group antigen; RhD polypeptide; RhDCw; RHDel; RHDVA(TT); Rhesus blood group CE and D; Rhesus blood group D antigen allele DIII type 7; rhesus D antigen; Rhesus system D polypeptide; RhII; RhK562-II; Rhl1; Rh-like polypeptide; RhPI; RHPII; RHXIII; truncated rhesus blood group D antigen; truncated rhesus D
Gene Symbols RHD
Host Species Mouse
Immunogen Recombinant Human RHD protein, Ser2-Gly87 (Accession #Q02161).
Purification Method Protein A/G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6007
Target Species Human, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.