Learn More
Invitrogen™ RHD Polyclonal Antibody

Description
Immunogen sequence: LGNLRMVISN IFNTDYHMNM MHIYVFAAYF GLSVAWCLPK PLPEGTEDKD QTATIPSLSA MLGALFLWMF WPSFNSALLR SPIERKNAVF NTYYAVAVSV V.
Specifications
Specifications
| Antigen | RHD |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.13 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | RHD |
| Gene Accession No. | O88298, Q8CF94 |
| Gene Alias | blood group antigen D; blood group protein RHD; blood group Rh(D) polypeptide; CD240D; D antigen (DCS); DIIIc; Erythrocyte membrane glycoprotein Rh30; Rh; Rh blood group antigen Evans; Rh blood group D antigen; Rh blood group protein; Rh blood group, D anitgen; Rh blood group, D antigen; RH polypeptide 2; Rh30; Rh4; Rhced; RHD; RhD blood group antigen; RhD polypeptide; RhDCw; RHDel; RHDVA(TT); Rhesus blood group CE and D; Rhesus blood group D antigen allele DIII type 7; rhesus D antigen; Rhesus system D polypeptide; RhII; RhK562-II; Rhl1; Rh-like polypeptide; RhPI; RHPII; RHXIII; truncated rhesus blood group D antigen; truncated rhesus D |
| Gene Symbols | RHD |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.