Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rheb Antibody (7G7), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | Rheb |
---|---|
Clone | 7G7 |
Dilution | ELISA, Immunocytochemistry/Immunofluorescence 1:20-1:200 |
Applications | ELISA, Immunocytochemistry |
Classification | Monoclonal |
Description
Rheb Monoclonal antibody specifically detects Rheb in Human samples. It is validated for ELISA, ImmunocytochemistrySpecifications
Rheb | |
ELISA, Immunocytochemistry/Immunofluorescence 1:20-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
GTP-binding protein Rheb, MGC111559, Ras homolog enriched in brainRHEB2Ras homolog enriched in brain 2 | |
A synthesized peptide derived from Human Rheb [UniProt Q15382] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
7G7 | |
ELISA, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Cancer, mTOR Pathway, Signal Transduction | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
6009 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title