Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rheb Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP176432
Description
Rheb Polyclonal specifically detects Rheb in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.Specifications
Rheb | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
GTP-binding protein Rheb, MGC111559, Ras homolog enriched in brainRHEB2Ras homolog enriched in brain 2 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Post-translationally modified Rheb is sometimes observed at higher molecular weight. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, ELISA | |
1 mg/mL | |
Western Blot 2-4 μg/mL, ELISA 1:100-1:2000 | |
AAH16155 | |
RHEB | |
Antibody was raised against a 14 amino acid synthetic peptide from the amino terminus of human Rheb. The immunogen is located within the first 50 amino acids of Rheb. Amino Acid Squence: PQSKSRKIAILGYR | |
0.1 mg | |
Cancer, mTOR Pathway, Signal Transduction | |
6009 | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction