Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rheb Antibody (Rheb 5D5), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250273B
Description
Rheb Monoclonal specifically detects Rheb in Human, Mouse samples. It is validated for Western Blot.Specifications
Rheb | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
RHEB | |
Human Rheb GST fusion | |
0.1 mL | |
Cancer, mTOR Pathway, Signal Transduction | |
6009 | |
Store at 4C in the dark. | |
IgG2a |
Western Blot | |
Rheb 5D5 | |
Western Blot | |
GTP-binding protein Rheb, MGC111559, Ras homolog enriched in brainRHEB2Ras homolog enriched in brain 2 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction