Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RHEB Mouse anti-Human, Mouse, Clone: 2F9H8, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | RHEB |
---|---|
Clone | 2F9H8 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-7129
![]() ![]() |
Proteintech Group Inc
667141IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Rheb encodes a member of the small GTPase superfamily and encodes a lipid-anchored, cell membrane protein with five repeats of the RAS-related GTP-binding region. This protein is vital in regulation of growth and cell cycle progression due to its role in the insulin/TOR/S6K signaling pathway. The protein has GTPase activity and shuttles between a GDP-bound form and a GTP-bound form, and farnesylation of the protein is required for this activity. Three pseudogenes have been mapped, two on chromosome 10 and one on chromosome 22.Specifications
RHEB | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
GTP binding protein Rheb, Ras homolog enriched in brain, RHEB, RHEB2 | |
RHEB | |
IgG1 | |
Protein G | |
Antibody |
2F9H8 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse | |
Q15382, Q921J2 | |
19744, 6009 | |
RHEB Fusion Protein Ag8887 | |
Primary | |
-20°C | |
RHEB |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title