Learn More
Invitrogen™ RHEB Polyclonal Antibody

Description
Immunogen sequence: MPQSKSRKIA ILGYRSVGKS SLTIQFVEGQ FVDSYDPTIE NTFTKLITVN GQEYHLQLVD TAGQDEYSIF PQTYSIDING YILVYSVTSI KSFEVIKVIH GKLLDMVGKV QIPIMLVGNK KDLHMERVIS YEEGKALAES WNAAFLESSA KENQTAVDVF RRIILEAEKM DGAASQGKSS.
Specifications
Specifications
| Antigen | RHEB |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.45 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 |
| Gene | RHEB |
| Gene Accession No. | Q15382, Q921J2 |
| Gene Alias | GTP-binding protein Rheb; Ras homolog enriched in brain; Ras homolog enriched in brain 2; RAS-homolog enriched in brain; RHEB; Rheb1; RHEB2 |
| Gene Symbols | RHEB |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.