Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RHEBL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562732
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562732 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562732 Supplier Invitrogen™ Supplier No. PA562732
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MPLVRYRKVV ILGYRCVGKT SLAHQFVEGE FSEGYDPTVE NTYSKIVTLG KDEFHLHLV Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 92%.

RhebL1 (ras homolog enriched in brain-like protein 1), also known as Rheb2 or GTPase RhebL1, is a 183 amino acid protein that belongs to the small GTPase superfamily and Rheb family. Localizing to the cell membrane as well as the cytoplasm, RhebL1 is ubiquitously expressed and is increased two-fold in many tumor cell lines. RhebL1 exhibits GTPase activity and may activate NF-kappa-B-mediated gene transcription. Regulating the activity of Rictor, RhebL1 also promotes signal transduction. RhebL1 exists as two alternatively spliced isoforms and is encoded by a gene that maps to human chromosome 12q13.12 and mouse chromosome 15 F1. Human chromosome 12 encodes over 1,100 genes and comprises approximately 4.5% of the human genome. Chromosome 12 is associated with a variety of diseases and afflictions, including hypochondrogenesis, achondrogenesis, Kniest dysplasia, Noonan syndrome and trisomy 12p, which causes facial developmental defects and seizure disorders. Binds GTP and exhibits intrinsic GTPase activity. May activate NF-kappa-B-mediated gene transcription. Promotes signal transduction through MTOR, activates RPS6KB1, and is a downstream target of the small GTPase-activating proteins TSC1 and TSC2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RHEBL1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RHEBL1
Gene Accession No. Q8TAI7
Gene Alias 1810036J22Rik; GTPase RhebL1; ras GTPase; Ras homolog enriched in brain like 1; Ras homolog enriched in brain like 1 c; ras homolog enriched in brain like-1 c; Ras homolog enriched in brain-like 1; ras homolog enriched in brain-like protein 1; RHEB like 1; Rheb2; RHEBL1; RHEBL1c; rheb-like protein 1
Gene Symbols RHEBL1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RHEBL1. Recombinant protein control fragment (Product #RP-97807).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 121268
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.