Learn More
Thermo Scientific™ RheFlex™ High-Pressure Fittings
Description
Ensure reliable high-pressure connections by using Thermo Scientific™ RheFlex™ High-Pressure Fittings. These fittings are precision machined from 316 stainless steel to be burr- and contaminant-free.
Specifications
Specifications
| Type | High Pressure SST Ferrule |
| For Use With (Equipment) | Model 8125 Injectors Only |
| Description | Ferrule; 0.5 mm for Model 8125 only |
| Quantity | 10 Pack |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.