Learn More
Thermo Scientific™ RheFlex™ High-Pressure Fittings
Catalog No. 03251810
Change view
Click to view available options
Type:
Extra Long High Pressure Fittings Set
High Pressure 1/16 in. Ferrule
High Pressure SST Ferrule
Long High Pressure Fittings Set
Short High Pressure Fittings Set
Quantity:
10 Pack
10/Pk.
5/Pk.
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Quantity
03-251-810 Long High Pressure Fittings Set 5/Pk.
03-251-813 High Pressure 1/16 in. Ferrule 10/Pk.
03-170-326 Extra Long High Pressure Fittings Set 10/Pk.
03-251-809 High Pressure 1/16 in. Ferrule 5/Pk.
03-251-812 Short High Pressure Fittings Set 10/Pk.
03-052-367 High Pressure SST Ferrule 10 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 03-251-810 Supplier Thermo Scientific™ Supplier No. 6000111
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Ensure reliable high-pressure connections with Thermo Scientific™ RheFlex™ High-Pressure Fittings.
Ensure reliable high-pressure connections by using Thermo Scientific™ RheFlex™ High-Pressure Fittings. These fittings are precision machined from 316 stainless steel to be burr- and contaminant-free.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) LC Columns
Material Stainless Steel
Type Long High Pressure Fittings Set
Description 10-32, 1/16 in. Long Fittings Set
Quantity 5/Pk.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.