Learn More
Thermo Scientific™ RheFlex™ Two-Piece PEEK Fittings
Catalog No. 03251801
Change view
Click to view available options
Type:
Fitting Set, Short
Fitting Set, Standard Length
Fitting Set, X-Long
Replacement Ferrule
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
03-251-801 Fitting Set, Standard Length
03-251-800 Replacement Ferrule
03-251-802 Fitting Set, Short
03-251-803 Fitting Set, X-Long
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 03-251-801 Supplier Thermo Scientific™ Supplier No. 6000054
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Thermo Scientific™ RheFlex™ Two-Piece PEEK Fittings provide inert, metal-free connections.
Thermo Scientific™ RheFlex™ Two-Piece PEEK Fittings provide inert, metal-free connections.
  • Slotted back-side of the ferrule is squeezed down onto the tube by the mating conical surface of the nut
  • May be used on 1/16 in. metal or plastic tubing reliably up to 5000 psi
  • Reusable ferrule and nut
TRUSTED_SUSTAINABILITY
Specifications
Material PEEK
For Use With (Equipment) PEEK Tubing
Description 1/16 in. Standard fitting set; 5/Pk.
Type Fitting Set, Standard Length
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.