Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Rhenium

Catalog No. GF02183217 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
GF02183217 1 Ea.
1 options

Catalog No. GF02183217

Supplier: Sigma Aldrich GF021832171EA

Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Rhenium

Specifications

CAS 7440-15-5
Melting Point 3180°C
Density 21.02 g/cm3 (literature)
Boiling Point 5596°C; 5627°C
Molecular Formula Re
Linear Formula Re
MDL Number MFCD00011195
Quantity 1 Ea.
Synonym Rhenium; RE005110
Molecular Weight (g/mol) 186.21
Percent Purity 99.97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.