Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rhenium pentacarbonyl chloride, 98%

Catalog No. p-190742
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

CAS: 14099-01-5 | C5ClO5Re | 361.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14099-01-5
Molecular Formula C5ClO5Re
Molecular Weight (g/mol) 361.71
MDL Number MFCD00013296
InChI Key JQUUAHKBIXPQAP-UHFFFAOYSA-M
Synonym pentacarbonylchlororhenium, carbon monoxide; chlororhenium, pentakis carbon monoxide ; chlororhenium, 5co.clre, rhenium i pentacarbonyl chloride, pentacarbonylchlororhenium i
PubChem CID 6096982
IUPAC Name carbon monoxide;chlororhenium
SMILES [C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].Cl[Re]

Specifications

Color White
Elemental Analysis 97.5% min.
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 361.71
Percent Purity 98%
Physical Form Fine Powder
Chemical Name or Material Rhenium pentacarbonyl chloride, 98%
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 237-948-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.