Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rhenium(VII) oxide, 99.995% (metals basis)

Catalog No. p-4824906
Click to view available options
Quantity:
2 g
10 g

CAS: 1314-68-7 | O7Re2 | 484.407 g/mol

Rhenium(VII) oxide is a precursor to organorhenium oxides which catalyze olefin oxidation, metathesis and other transformations. It is also used as a catalyst for hemiacetal substitution and as an oxidant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-68-7
Molecular Formula O7Re2
Molecular Weight (g/mol) 484.407
MDL Number MFCD00011199
InChI Key NBGOSNNAAHDRLK-UHFFFAOYSA-N
Synonym rhenium heptoxide, rhenium vii oxide, re2o7, dirhenium heptoxide, dirhenium heptaoxide, rhenium oxide re2o7, trioxo-trioxorheniooxy rhenium, trioxo trioxorheniooxy rhenium, rhenium vii oxide-re, rhenium vii oxide trace metals basis
PubChem CID 123106
IUPAC Name trioxo(trioxorheniooxy)rhenium
SMILES O=[Re](=O)(=O)O[Re](=O)(=O)=O

Specifications

Boiling Point 360°C
Melting Point 297°C
Color Yellow
Physical Form Crystalline
Quantity 2 g
Solubility Information Soluble in water and alcohol.
Formula Weight 484.4
Odor Odorless
Sensitivity Moisture sensitive
Density 6.103
Chemical Name or Material Rhenium(VII) oxide
Show More Show Less
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation DOT Class: 5.1, Packaging Group: III
EINECSNumber 215-241-9
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.