Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rhenium(VII) oxide, 99.995% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 4553809

Quantity 10g

 View more versions of this product

Catalog No. AA4553809


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

1314-68-7
O7Re2
MFCD00011199
Crystals
484.4
Rhenium(VII) oxide

Specifications

1314-68-7
O7Re2
MFCD00011199
Yellow
Rhenium(VII) oxide
Crystals
484.4
Odorless
10g

Safety and Handling

SDS & Safety Documents

H314-H318-H272: Causes severe skin burns and eye damage. Causes serious eye damage. May intensify fire; oxidizer.

DOTInformation : DOT Class: 5.1, Packaging Group: III

EINECSNumber : 215-241-9

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit