Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rhesus Macaque TNF-alpha Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $526.50
Specifications
Antigen | TNF-alpha |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
TNF-alpha Polyclonal specifically detects TNF-alpha in Rhesus Macaque samples. It is validated for Western Blot.Specifications
TNF-alpha | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Rhesus Macaque | |
APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha | |
TNF | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
P48094 | |
7124 | |
E. coli-derived recombinant rhesus macaque TNF-alpha Val77-Leu233 Accession # P48094 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects rhesus macaque TNF-alpha in direct ELISAs and Western blots. In Western blots, approximately 20% cross-reactivity with recombinant mouse TNF-alpha , recombinant rat TNF-alpha , and recombinant cotton rat TNF-alpha is observed, and less than 1% cross-reactivity with recombinant porcine TNF-alpha , recombinant human (rh) TRANCE, rhTWEAK, rhAPRIL, rhGITR Ligand, rhFas Ligand, and rhOX40 Ligand is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title