Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rhesus Macaque TNF-alpha Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB1070
This item is not returnable.
View return policy
Description
TNF-alpha Monoclonal specifically detects TNF-alpha in Rhesus Macaque samples. It is validated for Western Blot.Specifications
| TNF-alpha | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P48094 | |
| TNF | |
| E. coli-derived recombinant rhesus macaque TNF-alpha Val77-Leu233 Accession # P48094 | |
| 500 μg | |
| Primary | |
| Detects rhesus macaque TNF-alpha in direct ELISAs and Western blots. In direct ELISAs, aproximately 100% cross-rectivity with recombinant human TNF-alpha is observed and 20% cross-reactivity with TNF-alpha from mouse, rat, or cotton rat is observed. Addionally, in direct ELISAs, no cross-reactivity with recombinant human (rh) APRIL, rhBAFF, recombinant mouse (rm) BAFF R, rhEDA-A2, rmEDA, rhFas Ligand, rmFas Ligand, rhGITR Ligand, rhLIGHT, rhLT alpha 1 beta 2, rhLT alpha 2 beta 1, rhOX40 Ligand, rmOX40 Ligand, recombinant porcine TNF-alpha , rhTRAIL, rmTRAIL, rhTRANCE, rhTWEAK, rmTWEAK, or rhVEGI is observed. | |
| Rhesus Macaque | |
| Purified |
| Western Blot | |
| 182303 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7124 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction