Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rhesus Macaque TNF-alpha Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB10701
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB10701 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB10701 Supplier R&D Systems Supplier No. MAB10701
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

TNF-alpha Monoclonal specifically detects TNF-alpha in Rhesus Macaque samples. It is validated for Neutralization.

Specifications

Antigen TNF-alpha
Applications Neutralization
Classification Monoclonal
Clone 182309
Concentration LYOPH
Conjugate Unconjugated
Dilution Neutralization 0.2-0.8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P48094
Gene Alias APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha
Gene Symbols TNF
Host Species Rat
Immunogen E. coli-derived recombinant rhesus macaque TNF-alpha Val77-Leu233 Accession # P48094
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7124
Test Specificity Detects rhesus macaque TNF-alpha in direct ELISAs and Western blots. In direct ELISAs, 10-25% cross-reactivity with recombinant human (rh) TNF-alpha and recombinant porcine TNF-alpha is observed and no cross-reactivity with rhEDA-A2, recombinant mouse (rm) EDA, rhAPRIL, rhBAFF, rhFas L, rhGITR L, rhLIGHT, rhLymphotoxin alpha 1 beta 2, rhLymphotoxin alpha 2 beta 1, rhOX40 L, recombinant cotton rat TNF-alpha , rmTNF-alpha , recombinant rat TNF-alpha , rhTRAIL, rhTRANCE, and rhTWEAK is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Rhesus Macaque
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.