Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rheumatoid Factor Monoclonal Antibody (RF2-17-109.2), Invitrogen™
GREENER_CHOICE

Catalog No. 01671545
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-671-545 500 μg
PIMA545978 100 μg
PIMA545987 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-671-545 Supplier Invitrogen™ Supplier No. MA110592
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody reacts with 30% of IgM-RF from unrelated individuals. It does not recognize IgM, IgA, or IgG proteins lacking RF nor with F(ab’)2 fragments.

Antibodies directed against the Fc fragment of immunoglobulin G (IgG) are called rheumatoid factors (RFs). They are heterogenous and are usually IgM antibodies. RFs are used as a marker in individuals with suspected rheumatoid arthritis (RA) or other autoimmune conditions. The normal reference range for RF is less than 15 IU/mL.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rheumatoid Factor
Applications ELISA
Classification Monoclonal
Clone RF2-17-109.2
Concentration 1.8 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias RF
Host Species Mouse
Immunogen Purified proteins from patients with IgM cryoglobulins.
Purification Method Protein G
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.