Learn More
Invitrogen™ Rheumatoid Factor Monoclonal Antibody (RF2-17-109.2), Invitrogen™
Description
This antibody reacts with 30% of IgM-RF from unrelated individuals. It does not recognize IgM, IgA, or IgG proteins lacking RF nor with F(ab’)2 fragments.
Specifications
Specifications
| Antigen | Rheumatoid Factor |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | RF2-17-109.2 |
| Concentration | 1.8 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene Alias | RF |
| Host Species | Mouse |
| Immunogen | Purified proteins from patients with IgM cryoglobulins. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.