Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rhinovirus VP3 Monoclonal Antibody (L11A)
GREENER_CHOICE

Catalog No. PIMA518248
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA518248 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA518248 Supplier Invitrogen™ Supplier No. MA518248
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody recognizes the VP3 of numerous Rhino types (6) and does not react with Enteroviruses (5), Influenza A, Influenza B, RSV, Adenovirus, Para 1, Para 2, Para 3, Chlamydia pneumoniae, Mycoplasma pneumoniae or Hepatitis A viruses. Purity is >90%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Rhinovirus VP3
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone L11A
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias rotavirus gastroenteritis
Host Species Mouse
Immunogen Rhinovirus recombinant protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.