Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rho (-A, -B, -C), Rabbit, Unlabeled, Polyclonal,

Catalog No. ABS1848MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABS1848MI 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS1848MI Supplier MilliporeSigma™ Supplier No. ABS1848
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Rac1, RhoA, and Cdc42 (UniProt P63000, P61586, and P60953, respectively) are among the most studied Rho family of small GTPases that are involved in a wide variety of cellular functions such as cell polarity, vesicular trafficking, and cell cycle by regulating intracellular actin dynamics. In mammals, the Rho family contains 22 members which can be further divided into the Rac subfamily (Rac1, Rac2, Rac3, and RhoG), Cdc42 subfamily (Cdc42, TC10, TCL, Chip, and Wrch-1), RhoA subfamlly (RhoA, RhoB, and RhoC) and other Rho-GTPases (RhoE/Rnd3, RhoH/TTF, Rif, RhoBTB1, RhoBTB2, Miro-1, Miro-2, RhoD, Rnd1, and Rnd2).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rho (-A, -B, -C)
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene RHOA, ARH12, ARHA, RHO12, RHOB, ARH6, ARHB, RHOC, ARH9, ARHC
Gene Accession No. P61586;P62745;P08134
Host Species Rabbit
Immunogen KLH-conjugated synthetic peptide (SYPDTDVILMCFSIDSPDSLEN-KK) corresponding to amino acids 73-94 of human RhoA & RhoB, with two C-terminal lysines added to increase peptide solubility corresponding to human Rho (-A, -B, -C).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_001655/NP_004031/NP_786886
Test Specificity This polyclonal antibody is raised against an internal sequence highly conserved among RhoA (100%), RhoB (95%), and RhoC (100%).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.