Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RHO Rabbit anti-Human, Unconjugated, Clone: AADI-18, Abnova™
Rabbit Monoclonal Antibody.
Supplier: Abnova Corporation MAB20675
Description
RHO Monoclonal antibody specifically detects RHO in Human samples. It is validated for IHC, WB-Ti.Specifications
RHO | |
Monoclonal | |
Unconjugated | |
RHO | |
Rabbit | |
100 μg | |
6010 | |
Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing. | |
IgG |
Immunohistochemistry, Western Blot | |
AADI-18 | |
rhodopsin | |
CSNBAD1, MGC138309, MGC138311, OPN2, RP4 | |
A synthetic peptide corresponding to human RHO | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction