Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RHO Mouse anti-Bovine, Human, Clone: A531, Abnova™
Mouse monoclonal antibody raised against native bovine RHO.
Supplier: Abnova Corporation MAB12503
Specifications
RHO | |
Monoclonal | |
Unconjugated | |
Immunofluorescence (1:1000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot (1:5000) The optimal working dilution should be determined by the end user. | |
RHO | |
Native purified RHO from bovine retinal outer segments. | |
50 μL | |
Primary | |
Bovine, Human | |
Liquid |
Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
A531 | |
Mouse monoclonal antibody raised against native bovine RHO. | |
In PBS (10mM sodium azide) | |
Mouse | |
Protein G purification | |
RUO | |
509933 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction