Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rho Pan Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101028
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101028 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101028 Supplier Invitrogen™ Supplier No. PA5101028
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Rho A + B + C.

This gene encodes a member of the Rho family of small GTPases, which cycle between inactive GDP-bound and active GTP-bound states and function as molecular switches in signal transduction cascades. Rho proteins promote reorganization of the actin cytoskeleton and regulate cell shape, attachment, and motility. The protein encoded by this gene is prenylated at its C-terminus, and localizes to the cytoplasm and plasma membrane. It is thought to be important in cell locomotion. Overexpression of this gene is associated with tumor cell proliferation and metastasis. Multiple alternatively spliced variants, encoding the same protein, have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rho Pan
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RHOC
Gene Accession No. P08134, P17081, P61586, P61589, P62745, P62746, P62747, Q62159, Q8BYP3, Q8R527, Q9HBH0, Q9JJL4, Q9QUI0
Gene Alias AA017882; AI324259; AI845056; Aplysia ras-related homolog 12; Aplysia RAS-related homolog 6; aplysia ras-related homolog 9 (RhoC); aplysia ras-related homolog A; aplysia ras-related homolog A1; aplysia ras-related homolog A2; aplysia ras-related homolog B; ARH12; ARH6; ARH9; ARHA; Arha1; Arha2; Arhb; ARHC; ARHF; ARHQ; AV026554; epididymis secretory protein Li 42; h12; h6; H9; HEL-S-42; Ifld1; induced in fatty liver dystrophy 1; MST081; MSTP081; oncogene RHO H12; oncogene RHO H6; oncogene RHO H9; plysia ras-related homolog A2; Ras family member A; ras homolog 9 (RhoC); ras homolog A1; ras homolog A2; ras homolog B; ras homolog family member A; ras homolog family member B; ras homolog family member C; ras homolog family member F (in filopodia); ras homolog family member F, filopodia associated; ras homolog family member Q; ras homolog gene family, member A; ras homolog gene family, member A1; ras homolog gene family, member A2; ras homolog gene family, member AB; ras homolog gene family, member B; ras homolog gene family, member C; ras homolog gene family, member f; ras homolog gene family, member F (in filopodia); ras homolog gene family, member Q; RASL7A; ras-like protein family member 7A; Ras-like protein TC10; RAS-like, family 7, member A; ras-related GTP-binding protein TC10; RAS-related homolog 9; Rho cDNA clone 12; Rho cDNA clone 6; rho cDNA clone 9; rho family; Rho family GTPase; rho family GTPase Rif; Rho in filopodia; RHO12; Rhoa; Rhob; RHOC; rhoC GTPase; RHOF; RHOH12; RHOH6; RHOH9; Rhoq; rho-related GTP-binding protein RhoB; Rho-related GTP-binding protein RhoC; rho-related GTP-binding protein RhoF; Rho-related GTP-binding protein RhoQ; RIF; SIG-61; Silica-induced gene 61 protein; small GTP binding protein RhoA; small GTP binding protein RhoC; small GTPase Tc10; TC10; TC10A; Transforming protein RhoA
Gene Symbols RHOA, RHOB, RHOC, RHOF, RHOQ
Host Species Rabbit
Immunogen A synthesized peptide derived from human RHOC(Accession P08134), corresponding to amino acid residues K162-L193.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 104215, 117273, 11848, 11852, 11853, 23433, 23912, 295342, 387, 388, 389, 54509, 64373, 690130, 85428
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.