Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rho Polyclonal Antibody, Abbomax

Catalog No. 502203511
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-3511 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-3511 Supplier Abbomax Inc Supplier No. 50010054
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: MCF7 Cell Lysate Cellular location: Cell membrane, Cytoplasm.

Rho Polyclonal antibody specifically detects Rho in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen Rho
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene RHOQ
Gene Accession No. P08134, P17081, P61586, P61589, P62745, P62746, P62747, Q62159, Q8BYP3, Q8R527, Q9HBH0, Q9JJL4, Q9QUI0
Gene Alias AA017882; AI324259; AI845056; Aplysia ras-related homolog 12; Aplysia RAS-related homolog 6; aplysia ras-related homolog 9 (RhoC); aplysia ras-related homolog A; aplysia ras-related homolog A1; aplysia ras-related homolog A2; aplysia ras-related homolog B; ARH12; ARH6; ARH9; ARHA; Arha1; Arha2; Arhb; ARHC; ARHF; ARHQ; AV026554; epididymis secretory protein Li 42; h12; h6; H9; HEL-S-42; Ifld1; induced in fatty liver dystrophy 1; MST081; MSTP081; oncogene RHO H12; oncogene RHO H6; oncogene RHO H9; plysia ras-related homolog A2; Ras family member A; ras homolog 9 (RhoC); ras homolog A1; ras homolog A2; ras homolog B; ras homolog family member A; ras homolog family member B; ras homolog family member C; ras homolog family member F (in filopodia); ras homolog family member F, filopodia associated; ras homolog family member Q; ras homolog gene family, member A; ras homolog gene family, member A1; ras homolog gene family, member A2; ras homolog gene family, member AB; ras homolog gene family, member B; ras homolog gene family, member C; ras homolog gene family, member f; ras homolog gene family, member F (in filopodia); ras homolog gene family, member Q; RASL7A; ras-like protein family member 7A; Ras-like protein TC10; RAS-like, family 7, member A; ras-related GTP-binding protein TC10; RAS-related homolog 9; Rho cDNA clone 12; Rho cDNA clone 6; rho cDNA clone 9; rho family; Rho family GTPase; rho family GTPase Rif; Rho in filopodia; RHO12; Rhoa; Rhob; RHOC; rhoC GTPase; RHOF; RHOH12; RHOH6; RHOH9; Rhoq; rho-related GTP-binding protein RhoB; Rho-related GTP-binding protein RhoC; rho-related GTP-binding protein RhoF; Rho-related GTP-binding protein RhoQ; RIF; SIG-61; Silica-induced gene 61 protein; small GTP binding protein RhoA; small GTP binding protein RhoC; small GTPase Tc10; TC10; TC10A; Transforming protein RhoA
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the C-terminus human Rho protein
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 104215, 117273, 11848, 11852, 11853, 23433, 23912, 295342, 387, 388, 389, 54509, 64373, 690130, 85428
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.