Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RhoA Antibody (1A11-4G10), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222528
Description
RhoA Monoclonal specifically detects RhoA in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
RhoA | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500 - 1:1500, Immunohistochemistry 1:20 - 1:200, Immunocytochemistry/Immunofluorescence 1:10 - 1:200, Immunoprecipitation 2 ug, Immunohistochemistry-Paraffin 1:20 - 1:200 | |
h12, oncogene RHO H12, ras homolog gene family, member A, Rho cDNA clone 12, Rho12, RhoA, RHOH12ARH12ARHAAplysia ras-related homolog 12, small GTP binding protein RhoA, transforming protein RhoA | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
This antibody reacts with Human, Mouse and Rat. | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
1A11-4G10 | |
Unconjugated | |
P61586 | |
RHOA | |
Full-length recombinant human Rho A | |
100 μg | |
Cancer, mTOR Pathway, Signal Transduction, Wnt Signaling Pathway | |
387 | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction