Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RhoA Antibody (1C11), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
| Antigen | RhoA |
|---|---|
| Clone | 1C11 |
| Dilution | Western Blot 1:500-1:5000, ELISA |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
Description
RhoA Monoclonal antibody specifically detects RhoA in Human samples. It is validated for Western Blot, ELISASpecifications
| RhoA | |
| Western Blot 1:500-1:5000, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| h12, oncogene RHO H12, ras homolog gene family, member A, Rho cDNA clone 12, Rho12, RhoA, RHOH12ARH12ARHAAplysia ras-related homolog 12, small GTP binding protein RhoA, transforming protein RhoA | |
| A synthesized peptide derived from Human RHOA [UniProt P61586] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 1C11 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Cancer, mTOR Pathway, Phospho Specific, Signal Transduction, Wnt Signaling Pathway | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 387 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title