Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RHOA Mouse anti-Human, Mouse, Clone: 1B8-1C7, Abnova™

Catalog No. 89035555 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-555 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-555 Supplier Abnova Corporation Supplier No. MAB14157

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against full length recombinant human RHOA.

  • High specificity
  • High reactivity

Specifications

Antigen RHOA
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 1B8-1C7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant human RHOA.
Dilution Flow Cytometry (3-5 ug x 106 cells) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user.
Formulation In PBS (1 mg/mL BSA, 0.05% sodium azide).
Gene Accession No. P61586
Gene Alias ARH12/ARHA/RHO12/RHOH12
Gene Symbols RHOA
Host Species Mouse
Immunogen Recombinant His fusion protein corresponding to full length human RHOA.
Purification Method Protein A affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 387
Target Species Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.