Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RhoA/RhoB/RhoC Recombinant Rabbit Monoclonal Antibody (SR38-00)
GREENER_CHOICE

Catalog No. PIMA531993
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531993 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531993 Supplier Invitrogen™ Supplier No. MA531993
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

RhoA is described as a small GTPase protein that regulates the actin cytoskeleton in the formation of stress fibers. RhoA is also included in the related proteins of the Ras superfamily, which involves proteins in the regulation and timing of cell division.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RhoA/RhoB/RhoC
Applications Flow Cytometry, Western Blot, Immunocytochemistry, Western Blot, Western Blot
Classification Recombinant Monoclonal
Clone SR38-00
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Gene RHOC
Gene Accession No. P08134, P61586, P61589, P62745, P62746, P62747, Q62159, Q9QUI0
Gene Alias AA017882; AI324259; Aplysia ras-related homolog 12; Aplysia RAS-related homolog 6; aplysia ras-related homolog 9 (RhoC); aplysia ras-related homolog A; aplysia ras-related homolog A1; aplysia ras-related homolog A2; aplysia ras-related homolog B; ARH12; ARH6; Arh9; ARHA; Arha1; Arha2; ARHB; Arhc; h12; h6; h9; MST081; MSTP081; oncogene RHO H12; oncogene RHO H6; oncogene RHO H9; plysia ras-related homolog A2; Ras family member A; ras homolog 9 (RhoC); ras homolog A1; ras homolog A2; ras homolog B; ras homolog family member A; ras homolog family member B; ras homolog family member C; ras homolog gene family, member A; ras homolog gene family, member A1; ras homolog gene family, member A2; ras homolog gene family, member AB; ras homolog gene family, member B; ras homolog gene family, member C; RAS-related homolog 9; Rho; Rho cDNA clone 12; Rho cDNA clone 6; rho cDNA clone 9; Rho family GTPase; RHO12; Rhoa; Rhob; Rhoc; rhoC GTPase; RHOH12; RHOH6; RHOH9; rho-related GTP-binding protein RhoB; Rho-related GTP-binding protein RhoC; SIG-61; silica-induced gene 61 protein; small GTP binding protein RhoA; small GTP binding protein RhoC; transforming protein RhoA
Gene Symbols RHOA, RHOB, RHOC
Host Species Rabbit
Immunogen Recombinant protein within Human RhoC aa aa 1-193.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117273, 11848, 11852, 11853, 295342, 387, 388, 389, 64373
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.