Learn More
Invitrogen™ RhoB Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA588525
Description
Immunogen sequence: MAAIRKKLVV VGDGACGKTC LLIVFSKDEF PEVYVPTVFE NYVADIEVDG KQVELALWDT AGQEDYDRLR PLSYPDTDVI LMCFSVDSPD SLENIPEKWV PEVKHFCPNV PIILVANKKD LRSDEHVRTE LARMKQEPVR TDDGRAMAVR IQAYDYLECS AKTKEGVREV FETATRAALQ KRYGSQNGCI NCCKVL; Positive Samples: Mouse brain, Rat brain; Cellular Location: Cell membrane, Cleavage furrow, Late endosome membrane, Lipid-anchor, Nucleus.
Rho-related GTP-binding protein (RhoB) mediates apoptosis in neoplastically transformed cells after DNA damage. While not essential for development, it does affect cell adhesion and growth factor signaling in transformed cells. RhoB targets PKN1 to endosomes and is involved in trafficking of the EGF receptor from late endosomes to lysosomes. Also required for stability and nuclear trafficking of AKT1/AKT which promotes endothelial cell survival during vascular development. Serves as a tumor supressor - deletion of RhoB causes tumor formation.
Specifications
RhoB | |
Polyclonal | |
Unconjugated | |
RHOB | |
AA017882; Aplysia RAS-related homolog 6; aplysia ras-related homolog B; Arh6; Arhb; h6; MST081; MSTP081; oncogene RHO H6; ras homolog B; ras homolog family member B; ras homolog gene family, member AB; ras homolog gene family, member B; Rho; rho cDNA clone 6; rhoB; RHOH6; Rho-related GTP-binding protein RhoB | |
Rabbit | |
Affinity Chromatography | |
RUO | |
11852, 64373 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.26 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P62746, P62747 | |
RHOB | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-196 of human RhoB (NP_0040311). | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.