Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RhoB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115844
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115844 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115844 Supplier Invitrogen™ Supplier No. PA5115844
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total RHOB.

Rho-related GTP-binding protein (RhoB) mediates apoptosis in neoplastically transformed cells after DNA damage. While not essential for development, it does affect cell adhesion and growth factor signaling in transformed cells. RhoB targets PKN1 to endosomes and is involved in trafficking of the EGF receptor from late endosomes to lysosomes. Also required for stability and nuclear trafficking of AKT1/AKT which promotes endothelial cell survival during vascular development. Serves as a tumor supressor - deletion of RhoB causes tumor formation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RhoB
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene RHOB
Gene Accession No. P62745, P62746, P62747
Gene Alias AA017882; Aplysia RAS-related homolog 6; aplysia ras-related homolog B; Arh6; Arhb; h6; MST081; MSTP081; oncogene RHO H6; ras homolog B; ras homolog family member B; ras homolog gene family, member AB; ras homolog gene family, member B; Rho; rho cDNA clone 6; rhoB; RHOH6; Rho-related GTP-binding protein RhoB
Gene Symbols RHOB
Host Species Rabbit
Immunogen A synthesized peptide derived from human RHOB(Accession P62745), corresponding to amino acid residues K98-A148.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11852, 388, 64373
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.