Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RhoB Recombinant Rabbit Monoclonal Antibody (3B10K10)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA551248
Description
Immunogen sequence: RPLSYPDTDV ILMCFSVDSP DSLENIPEKW VPEVKHFCPN VPIILVANKK DLRSDEHVRT ELARMKQEPV RTDDGRAMAV R.
Rho-related GTP-binding protein (RhoB) mediates apoptosis in neoplastically transformed cells after DNA damage. While not essential for development, it does affect cell adhesion and growth factor signaling in transformed cells. RhoB targets PKN1 to endosomes and is involved in trafficking of the EGF receptor from late endosomes to lysosomes. Also required for stability and nuclear trafficking of AKT1/AKT which promotes endothelial cell survival during vascular development. Serves as a tumor supressor - deletion of RhoB causes tumor formation.
Specifications
RhoB | |
Recombinant Monoclonal | |
0.07 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
P62745, P62746, P62747 | |
RHOB | |
A synthetic peptide corresponding to a sequence within amino acids 70-150 of human RhoB (NP_004031.1). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
Liquid |
ELISA, Western Blot | |
3B10K10 | |
Unconjugated | |
RHOB | |
AA017882; Aplysia RAS-related homolog 6; aplysia ras-related homolog B; Arh6; Arhb; h6; MST081; MSTP081; oncogene RHO H6; ras homolog B; ras homolog family member B; ras homolog gene family, member AB; ras homolog gene family, member B; Rho; rho cDNA clone 6; rhoB; RHOH6; Rho-related GTP-binding protein RhoB | |
Rabbit | |
Affinity chromatography | |
RUO | |
11852, 388, 64373 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction