Learn More
Description
Used for: labeling the ECM in live tissues.
Application: fluorescent microscopy including 2P and STED microscopy. Enables wash-free visualization of ECM in vitro and in vivo and is suitable for long-term imaging studies.
Properties and Photophysical Data: Rhobo6 provides broad substrate profile, superior tissue penetration, and negligible photobleaching. Excitation and emission maxima (λ) measured in 2 M sorbitol to mimic binding, are 535 nm and 561 nm, respectively; quantum yield = 0.75; extinction coefficient = 97,500 M-1cm-1.
Please refer to this guide for further information on how to use this product.
Specifications
Specifications
| Chemical Name or Material | (E)-2-(6-((2-Boronobenzyl)amino)-3-((2-boronobenzyl)iminio)-3H-xanthen-9-yl)-4-carboxybenzoate trifluoroacetate |
| Quantity | 76 μg |
| Product Line | Tocris |
| Target | Live-cell Imaging |
| Molecular Formula | C35H28B2N2O9.CF3CO2H |
| Purity | 85% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
