Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RhoBTB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111119
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111119 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111119 Supplier Invitrogen™ Supplier No. PA5111119
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TQTLTGWSKG FIGMHREMQV NPISKRMGPM TVVRMDASVQ PGPFRTLLQF LYTGQLDEKE KDLVGLAQIA EVLEM Highest antigen sequence indentity to the following orthologs: Rat - 77%, Mouse - 76%.

RHOBTB1 (Rho-related BTB domain-containing protein 1) and RHOBTB3 (Rho-related BTB domain-containing protein 3) each contain two BTB (POZ) domains and belong to the RhoBTB subfamily of Rho GTPases. Members of the RhoBTB subfamily are evolutionarily conserved and are characterized by a proline-rich region, a GTPase domain and two tandem BTB repeats. While both RHOBTB1 and RHOBTB3 are expressed ubiquitously, RHOBTB1 is found at high levels in placenta, stomach, testis, kidney and skeletal muscle, whereas RHOBTB3 is found at high levels in neural and cardiac tissues. RHOBTB1 is thought to play a role in GTPase-mediated signaling and may participate in organization of the Actin filament system. Additionally, RHOBTB1 expression is decreased in head and neck carcinomas, suggesting a possible role for RHOBTB1 as a tumor suppressor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RhoBTB1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RHOBTB1
Gene Accession No. O94844
Gene Alias 1700008H16Rik; 3110048G13Rik; AI173445; AI573858; AV350930; DBC2; KIAA0717; KIAA0740; MGC33059; MGC33841; p83; Rho related BTB domain containing 1; Rhobtb1; RHOBTB2; Rho-related BTB domain containing 1; rho-related BTB domain-containing protein 1; zmp:0000000868
Gene Symbols RHOBTB1
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human RhoBTB1. Recombinant protein control fragment (Product #RP-97009).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9886
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.