Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RHOBTB2 Antibody (DBC2/3362), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308659AF700
Description
RHOBTB2 Monoclonal antibody specifically detects RHOBTB2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein ArraySpecifications
RHOBTB2 | |
Monoclonal | |
Alexa Fluor 700 | |
DBC2KIAA0717Deleted in breast cancer 2 gene protein, p83, Rho-related BTB domain containing 2, rho-related BTB domain-containing protein 2 | |
Human recombinant RHOBTB2 protein fragment (around aa554-604) (exact sequence is proprietary) (Uniprot: Q9BYZ6) | |
100 μg | |
Signal Transduction | |
23221 | |
Store at 4°C in the dark. | |
IgG2c λ |
Immunohistochemistry (Paraffin), Protein Array | |
DBC2/3362 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction