Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RhoBTB2 Monoclonal Antibody (2G6)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA521429
Description
Peptide Sequence: TISAHKPLLI SSCDWMAAMF GGPFVESSTR EVVFPYTSKS CMRAVLEYLY TGMFTSSPDL DDMKLIILAN RLCLPHLVAL TEQYTVTGLM EATQMMVDID GDVLVFLELA.
RHOBTB2 is a member of the evolutionarily conserved RHOBTB subfamily of Rho GTPases. For background information on RHOBTBs, see RHOBTB1.
Specifications
RhoBTB2 | |
Monoclonal | |
0.2-1.0 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q9BYZ6 | |
RHOBTB2 | |
RHOBTB2 (AAH34917, 510 a.a. ~ 619 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA | |
2G6 | |
Unconjugated | |
RHOBTB2 | |
Dbc2; Deleted in breast cancer 2 gene protein; deleted in breast cancer 2 gene protein homolog; E130206H14Rik; KIAA0717; KIAA0740; mKIAA0717; p83; Rho related BTB domain containing 2; Rho related BTB domain containing protein 2; RHOBTB1; RHOBTB2; Rho-related BTB domain containing 2; rho-related BTB domain-containing protein 2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
23221 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction