Learn More
Invitrogen™ RhoBTB3 Monoclonal Antibody (3A10)

Description
Peptide Sequence: MSIHIVALGN EGDTFHQDNR PSGLIRTYLG RSPLVSGDES SLLLNAASTV ARPVFTEYQA SAFGNVKLVV HDCPVWDIFD SDWYTSRNLI GGADIIVIK.
Specifications
Specifications
| Antigen | RhoBTB3 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 3A10 |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | RHOBTB3 |
| Gene Accession No. | O94955 |
| Gene Alias | 1700040C17Rik; 2610033K01Rik; 4930503C18Rik; AI317148; AW208826; Kiaa0878; mKIAA0878; Rho related BTB domain containing 3; RHOBTB 3; RHOBTB3; Rho-related BTB domain containing 3; rho-related BTB domain-containing protein 3 |
| Gene Symbols | RHOBTB3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.