Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RhoBTB3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598118
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598118 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598118 Supplier Invitrogen™ Supplier No. PA598118
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RhoBTB3 Polyclonal Antibody for Western Blot, ELISA

The Rho subfamily of Ras-related GTPases controls multiple aspects of cell function, including cytoskeletal rearrangement, nuclear signaling and cell growth. RHOBTB1 (Rho-related BTB domain-containing protein 1) and RHOBTB3 (Rho-related BTB domain-containing protein 3) each contain two BTB (POZ) domains and belong to the RhoBTB subfamily of Rho GTPases. Members of the RhoBTB subfamily are evolutionarily conserved and are characterized by a proline-rich region, a GTPase domain and two tandem BTB repeats. While both RHOBTB1 and RHOBTB3 are expressed ubiquitously, RHOBTB1 is found at high levels in placenta, stomach, testis, kidney and skeletal muscle, whereas RHOBTB3 is found at high levels in neural and cardiac tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RhoBTB3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene RHOBTB3
Gene Accession No. O94955
Gene Alias 1700040C17Rik; 2610033K01Rik; 4930503C18Rik; AI317148; AW208826; Kiaa0878; mKIAA0878; Rho related BTB domain containing 3; RHOBTB 3; RHOBTB3; Rho-related BTB domain containing 3; rho-related BTB domain-containing protein 3
Gene Symbols RHOBTB3
Host Species Rabbit
Immunogen Recombinant Human Rho-related BTB domain-containing protein 3 protein (251-500AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22836
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.