Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RhoC Antibody (OTI3A5), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273888FR
Description
RhoC Monoclonal specifically detects RhoC in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
RhoC | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
RHOC | |
Synthetic peptide: VFEMATRAGLQVRKNKRRRG, corresponding to C terminal of human RhoC. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Western Blot, Immunohistochemistry | |
OTI3A5 | |
Western Blot, Immunohistochemistry | |
ARH9ARHCRho cDNA clone 9, H9, MGC1448, MGC61427, oncogene RHO H9, ras homolog gene family, member C, RAS-related homolog 9, RhoC, rhoC GTPase, RHOH9, rho-related GTP-binding protein RhoC, small GTP binding protein RhoC | |
Mouse | |
Affinity Purified | |
Signal Transduction | |
389.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction