Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RhoC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA577866
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA577866 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA577866 Supplier Invitrogen™ Supplier No. PA577866
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: A431, mouse liver Predicted Reactivity: Rat (100%), Xenopus laevis (89%), Chicken (96%), Bovine (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

RhoC is part of blood-group antigens that are generally defined as molecules formed by sequential addition of saccharides to the carbohydrate side chains of lipids and proteins detected on erythrocytes and certain epithelial cells. The A, B and H antigens are reported to undergo modulation during malignant cellular transformation. Blood group related antigens represent a group of carbohydrate determinants carried on both glycolipids and glycoproteins. They are usually mucin-type, and are detected on erythrocytes, certain epithelial cells, and in secretions of certain individuals. Sixteen genetically and biosynthetically distinct but inter-related specificities belong to this group of antigens, including A, B, H, Lewis A, Lewis B, Lewis X, Lewis Y, and precursor type 1 chain antigens.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RhoC
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene Rhoc
Gene Accession No. P08134, Q62159
Gene Alias AI324259; aplysia ras-related homolog 9 (RhoC); AR ARHC; Arh9; ARHC; h9; MGC1448; MGC61427; oncogene RHO H9; ras homolog 9 (RhoC); ras homolog family member C; ras homolog gene family, member C; RAS-related homolog 9; Rho; rho cDNA clone 9; RHO MGC1448; RHOC; rhoC GTPase; RHOH9; Rho-related GTP-binding protein RhoC; SIG-61; silica-induced gene 61 protein; small GTP binding protein RhoC
Gene Symbols Rhoc
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human RhoC. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11853, 389
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.