Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RhoD Antibody (OTI2F7), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273889PEATT594
Description
RhoD Monoclonal specifically detects RhoD in Human samples. It is validated for Flow Cytometry.Specifications
RhoD | |
Monoclonal | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
RHOD | |
Full length recombinant protein of human RhoD (NP_055393) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG2b |
Flow Cytometry | |
OTI2F7 | |
Flow Cytometry | |
ARHDRHOM, ras homolog D, ras homolog gene family, member A, ras homolog gene family, member D, Rho, RhoD, RhoHP1, rho-related GTP-binding protein RhoD, Rho-related protein HP1 | |
Mouse | |
Affinity Purified | |
mTOR Pathway, Signal Transduction | |
29984.0 | |
Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction