Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RHOD Monoclonal Antibody (OTI2F7), TrueMAB™, OriGene
SDP

Catalog No. 502540652
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0652 30 μL
50-167-4672 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0652 Supplier OriGene Supplier No. TA503891S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RHOD Monoclonal antibody specifically detects RHOD in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot

Specifications

Antigen RHOD
Applications Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI2F7
Concentration 0.85 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene PDIA4
Gene Accession No. P08003, P13667
Gene Alias AI326383; aplysia ras-related homolog D; Arhd; LOW QUALITY PROTEIN: rho-related GTP-binding protein RhoD; ras homolog D; ras homolog D (RhoD); ras homolog family member D; ras homolog gene family, member A; ras homolog gene family, member D; Rh(o); Rh(o)ii; Rho; RHOD; RhoHP1; RHOM; Rho-related GTP-binding protein RhoD; Rho-related protein HP1
Host Species Mouse
Immunogen Full length human recombinant protein of human PDIA4 produce in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12304, 9601
Target Species Human, Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.