Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rhodamine B Monoclonal Antibody (1E7)
GREENER_CHOICE

Catalog No. PIMA549636
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549636 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549636 Supplier Invitrogen™ Supplier No. MA549636
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Antibody tested in indirect ELISA.

Rhodamine B is a fluorescent dye molecule that belongs to the rhodamine family. It is commonly used as a tracer molecule in various fields such as biology, chemistry, and materials science. Rhodamine B has a high quantum yield, meaning it emits strong fluorescence signals, and it is also relatively stable and easy to handle. Its bright and intense color, along with its excellent photostability and chemical stability, makes it an excellent choice for use in fluorescence microscopy, flow cytometry, and other imaging techniques.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rhodamine B
Applications ELISA, Immunohistochemistry, Lateral Flow, SDS-Page
Classification Monoclonal
Clone 1E7
Concentration 4.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Host Species Mouse
Immunogen Conjugate of RB hapten and KLH.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.