Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rhodamine B 95.0+%, TCI America™
SDP

Catalog No. p-7135671 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
250 g

Catalog No. R004025G

Supplier: TCI America R004025G

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 81-88-9
Molecular Formula C28H31ClN2O3
Molecular Weight (g/mol) 479.02
MDL Number MFCD00011931
InChI Key PYWVYCXTNDRMGF-UHFFFAOYSA-N
Synonym rhodamine b, basic violet 10, brilliant pink b, tetraethylrhodamine, rhodamine o, rheonine b, rhodamine fb, calcozine red bx, symulex magenta f, rhodamine
PubChem CID 6694
ChEBI CHEBI:52334
IUPAC Name 9-(2-carboxyphenyl)-3,6-bis(diethylamino)-10λ⁴-xanthen-10-ylium chloride
SMILES [Cl-].CCN(CC)C1=CC2=[O+]C3=CC(=CC=C3C(C3=CC=CC=C3C(O)=O)=C2C=C1)N(CC)CC

Specifications

Melting Point 211°C
Color Green-Yellow
Quantity 25 g
Formula Weight 479.02
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Rhodamine B
EINECSNumber (5)-1973&(5)-4056
RTECSNumber BP3675000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.