Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rhodium acetate, ca. 40% Rh, brown, water soluble

Catalog No. p-41353
Click to view available options
Quantity:
1 g
5 g

CAS: 42204-14-8 | C2H3O2Rh | 161.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42204-14-8
Molecular Formula C2H3O2Rh
Molecular Weight (g/mol) 161.95
MDL Number MFCD03410291
InChI Key NENDHUHGFRLXEN-UHFFFAOYSA-M
Synonym rhodium 3+ ion acetate
PubChem CID 3660445
IUPAC Name acetic acid;rhodium
SMILES [Rh].CC([O-])=O

Specifications

Color Brown
Physical Form Powder
Quantity 1 g
Linear Formula Rh(CO2CH3)x
Solubility Information Solubility in water: soluble
Formula Weight 130.96
Percent Purity ∼40% (Rh)
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Rhodium acetate, ca. 40% Rh
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Suspected of causing genetic defects.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical
EINECSNumber 255-707-9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.